Skip to Content
Merck
All Photos(1)

Key Documents

PZ0129

Sigma-Aldrich

CP-380736

≥98% (HPLC)

Synonym(s):

6,7-Bis(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one, 6,7-Bis(2-methoxyethoxy)-4(3H)-quinazolinone, PF-00520893

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$841.00
25 MG
R$2,673.00

R$841.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$841.00
25 MG
R$2,673.00

About This Item

Empirical Formula (Hill Notation):
C14H18N2O5
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

R$841.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

COCCOc1cc2N=CNC(=O)c2cc1OCCOC

InChI

1S/C14H18N2O5/c1-18-3-5-20-12-7-10-11(15-9-16-14(10)17)8-13(12)21-6-4-19-2/h7-9H,3-6H2,1-2H3,(H,15,16,17)

InChI key

PMQWTUWLIGJTQD-UHFFFAOYSA-N

Biochem/physiol Actions

CP-380736 is an inhibitor of the epidermal growth factor receptor (EGFR). EGFR is a tyrosine kinase that activates MAPK, JNK, and Akt pathways, and is an important mediator of several types of cancer, including lung cancer and glioblastoma multiforme.
CP-380736 is selective inhibitor of epidermal growth factor receptor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Belén Sánchez-Maldonado et al.
Stem cells and development, 27(22), 1557-1576 (2018-09-27)
Cell spheroids are inducible or spontaneously generated cell aggregates produced in vitro that can provide a valuable model for developmental biology, stem cell biology, and cancer therapy research. This investigation aimed to define the cellular identity of spheroids spontaneously generated

Articles

Our company offers many products related to EGFR for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service