Skip to Content
Merck
All Photos(4)

Key Documents

PRS3897

Sigma-Aldrich

Anti-TRPC6 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Htrp6, Anti-Short transient receptor potential channel 6, Anti-Transient receptor potential cation channel, subfamily C, member 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,731.00

R$3,731.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,731.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,731.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 100-102 kDa

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPC6(7225)

Immunogen

a 14 amino acid peptide from near the carboxy-terminus of human TRPC6.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3897.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyun Wang et al.
American journal of nephrology, 55(3), 345-360 (2024-02-09)
The aim of this study was to explore the renoprotective effects of Klotho on podocyte injury mediated by complement activation and autoantibodies in idiopathic membranous nephropathy (IMN). Rat passive Heymann nephritis (PHN) was induced as an IMN model. Urine protein
Cristina I Linde et al.
American journal of physiology. Heart and circulatory physiology, 303(7), H784-H794 (2012-07-31)
The mechanisms by which NaCl raises blood pressure (BP) in hypertension are unresolved, but much evidence indicates that endogenous ouabain is involved. In rodents, arterial smooth muscle cell (ASMC) Na(+) pumps with an α(2)-catalytic subunit (ouabain EC(50) ≤1.0 nM) are
Kimberly A Smith et al.
American journal of respiratory cell and molecular biology, 53(3), 355-367 (2015-01-09)
Hypoxic pulmonary vasoconstriction (HPV) is an important physiological response that optimizes the ventilation/perfusion ratio. Chronic hypoxia causes vascular remodeling, which is central to the pathogenesis of hypoxia-induced pulmonary hypertension (HPH). We have previously shown that Notch3 is up-regulated in HPH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service