Skip to Content
Merck
All Photos(4)

Documents

PRS3331

Sigma-Aldrich

Anti-XIAP antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-X-linked inhibitor of apoptosis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 55 kDa

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XIAP(331)

General description

XIAP (X-linked inhibitor of apoptosis protein) belongs to the inhibitor of apoptosis (IAP) family. It is located on chromosome Xq25.

Immunogen

a synthetic peptide corresponding to 13 amino acids at the C-terminus of human XIAP.

Application

Anti-XIAP has been used in immunohistochemistry {16,17}.

Biochem/physiol Actions

XIAP (X-linked inhibitor of apoptosis protein) modulates several cellular processes like TGF-β (transforming growth factor β) signaling, NF-κB (nuclear factor-κB) pathway, autophagy and inflammatory responses. XIAP mutations are associated with X-linked lymphoproliferative syndrome type 2 (XLP-2).

Linkage

The action of this antibody can be blocked using blocking peptide SBP3331.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distinct 5' UTRs regulate XIAP expression under normal growth conditions and during cellular stress
Riley A, et al.
Nucleic Acids Research, 38(14), 4665-4674 (2010)
Yiping Zhang et al.
OncoTargets and therapy, 11, 8751-8759 (2018-12-26)
Carboplatin is a first-line treatment for ovarian cancer. However, most patients develop resistance and undergo disease recurrence. This study aims to explore the relationship between the expression of X-linked inhibitor of apoptosis protein (XIAP) and carboplatin sensitivity in ovarian cancer.
Zhuan Zhou et al.
EBioMedicine, 15, 48-61 (2017-01-04)
The emerging regulatory role of deubiquitinases (DUBs) has been implicated in various fundamental processes and pathogenesis. To determine the pivotal role that DUBs play in mediating tumorigenesis, we have performed a non-biased screen of 67 human DUBs based on a
Chao Chen et al.
Oncology letters, 15(1), 821-832 (2018-02-07)
The X-linked inhibitor of apoptosis protein (XIAP) is the best characterized member of the IAP family and is a potent inhibitor of the caspase/apoptosis pathway. It has also been revealed that XIAP has additional biological functions that rely on its
Dilys T H Leung et al.
Molecular cancer therapeutics, 18(2), 364-375 (2018-12-12)
Ovarian granulosa cell tumors (GCT) are characterized by indolent growth and late relapse. No therapeutic modalities aside from surgery have proven effective. We previously reported overexpression of the nuclear receptor, peroxisome proliferator-activated receptor-gamma (PPARγ), and constitutive activity of the NFκB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service