PLA0196
Rabbit anti-STUB1/CHIP Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
CHIP, CLL-associated antigen KW-8, E3 ubiquitin protein ligase, HSPABP2, NY-CO-7, SCAR16, SDCCAG7, STIP1 homology and U box-containing protein 1, STIP1 homology and U-box containing protein 1, UBOX1, antigen NY-CO-7, carboxy terminus of Hsp70-interacting protein, heat shock protein A binding protein 2 (c-terminal), serologically defined colon cancer antigen 7
About This Item
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
species reactivity
mouse, human
technique(s)
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000
accession no.
NP_005852.2
shipped in
wet ice
storage temp.
2-8°C
Gene Information
rabbit ... STUB1/CHIP(10273)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service