Skip to Content
Merck
All Photos(2)

Key Documents

PLA0117

Sigma-Aldrich

Rabbit anti-RIF1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

FLJ10599, FLJ12870, RAP1 interacting factor homolog, RAP1 interacting factor homolog (yeast), rap1-interacting factor 1 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 1- 5 μg/mg

accession no.

NP_060621.3

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... RIF1(55183)

Immunogen

The epitope recognized by PLA0117 maps to a region between residues 1 and 50 of human Rap1 Interacting Factor 1 using the numbering given in entry NP_060621.3 (GeneID 55183).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

RAP1 interacting factor (RIF1) localizes to dysfunctional telomeres and functions in the S-phase checkpoint. In response to double-stranded breaks (DSBs), RIF1 functions in the ATM and ATR mediated response to DNA damage. RIF1 foci formation in response to DNA damage is completely dependent on ATM.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service