Skip to Content
Merck
All Photos(2)

Key Documents

PLA0087

Sigma-Aldrich

Rabbit anti-MAD2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

HSMAD2, MAD2, MAD2 (mitotic arrest deficient, MAD2-like protein 1, homolog)-like 1, homolog-like 1, mitotic arrest deficient, mitotic arrest deficient 2-like protein 1, yeast

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$1,578.00

R$1,578.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
100 μL
R$1,578.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,578.00


Estimated to ship onJune 02, 2025


biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

Q13257

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... MAD2(4085)

Immunogen

The epitope recognized by PLA0087 maps to a region between residue 150 and the C-terminus (residue 205) of human Mitotic Arrest Deficient 2 using the numbering given in SwissProt entry Q13257 (GeneID 4085).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

MAD2 (Mitotic Arrest Deficiency 2) is a component of the mitotic spindle-assembly checkpoint that ensures that all chromosomes are aligned at the metaphase plate before the onset of anaphase. MAD2 is a subunit of the MCC complex that includes BubR1, Bub3, Cdc20 and Mad2, which may function to inhibit the APC/c (Anaphase Promoting Complex/Cyclosome). MAD2 is regulated by MAD1.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service