Skip to Content
Merck
All Photos(1)

Documents

PLA0012

Sigma-Aldrich

Rabbit anti-Phospho SMC1 (S957) Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CDLS2, DXS423E, KIAA0178, SB1.8, SMC protein 1A, SMC-1-alpha, SMC-1A, SMC1, SMC1 (structural maintenance of chromosomes 1, SMC1 structural maintenance of chromosomes 1-like 1 (yeast), SMC1L1, SMC1alpha, Smcb, segregation of mitotic chromosomes 1, yeast)-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2- 6 μg/mg
western blot: 1:200-1:2,000

accession no.

NP_006297.2

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... SMC1A(8243)

Immunogen

Immunogen for PLA0012 was a phosphorylated synthetic peptide, which represented a portion of human Structural Maintenance of Chromosomes 1 (GeneID 8243) around serine 957 according to the numbering given in entry NP_006297.2.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Structural maintenance of chromosomes (SMC) proteins are part of the cohesin and condensin multisubunit complexes that play a critical role in chromosome organization, segregation, gene regulation, and DNA repair. The core cohesin complex includes a heterodimer of SMC1 and SMC3 that functions by forming a ring-like structure that holds sister chromatids together. Cohesin is important to sister chromatid separation and segregation during mitosis and meiosis and also plays a role in double-strand break repair by homologous recombination.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service