PIR01
Inhibitor Resistant Genotyping PCR ReadyMix™, Low ROX™
with Low ROX for ABI and Stratagene instruments
Synonym(s):
Inhibitor Resistant Genotyping PCR ReadyMix™
About This Item
Recommended Products
form
liquid
usage
sufficient for 1250 reactions
sufficient for 250 reactions
sufficient for 5000 reactions
feature
Difficult Templates/Specialty Enzymes PCR
dNTPs included
hotstart
storage condition
protect from light
technique(s)
qPCR: suitable
color
light blue
input
crude DNA
detection method
probe-based
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
A key component of Inhibitor Resistant Genotyping PCR ReadyMix is an ultra-pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This proprietary polymerase mix is highly resistant to PCR inhibitors and provides an extremely stringent automatic hotstart allowing reaction assembly, and temporary storage, at room temperature prior to PCR amplification.
Application
- Applied Biosystems 7500
- Applied Biosystems 7500 Fast
- Applied Biosystems ViiA 7
- Stratagene Mx3000P®
- Stratagene Mx3005P®
- Stratagene Mx4000™
Features and Benefits
- Assay results in as little as 33 minutes
- Highly efficient and sensitive real-time PCR results
- Little/no optimization required
- Compatible with both fast and conventional PCR cycling protocols
Components
250 reactions* = 2 X 1.25 mL tubes
1250 reactions* = 10 X 1.25 mL tubes
5000 reactions* = 1 X 50 mL tube
*number of reactions based on a 20uL volume
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service