Skip to Content
Merck
All Photos(1)

Key Documents

P9436

Sigma-Aldrich

Pectin, esterified potassium salt from citrus fruit

extent of labeling: 55-70% esterified

Synonym(s):

Pectic acid, potassium salt, Potassium pectate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

citrus

form

powder

extent of labeling

55-70% esterified

Looking for similar products? Visit Product Comparison Guide

General description

Pectin is a polysaccharide rich in galacturonic acid and is localized to the plant cell wall. It possesses a complex structure and function.

Application

Pectin, esterified potassium salt from citrus fruit has been used:
  • in enzyme linked immunosorbent assay (ELISA)
  • in the determination of optimum pH of pectate lyase 10A (Pel10Acm) activity and in high-performance anion-exchange chromatography (HPAEC) as a part of Pel10Acm assays
  • to compare its gel properties with that of pectin extracted from sunflower head residues

Biochem/physiol Actions

Pectin is essential for plant development including growth, morphological changes and immunity. It serves as a stabilizing polymer and is useful in food and speciality products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell wall pectins and xyloglucans are internalized into dividing root cells and accumulate within cell plates during cytokinesis
Baluvska F, et al.
Protoplasma, 225(3-4), 141-155 (2005)
Pectate lyase 10A from Pseudomonas cellulosa is a modular enzyme containing a family 2a carbohydrate-binding module
Brown IE, et al.
The Biochemical Journal, 355(1), 155-165 (2001)
Extraction and characterization of sunflower pectin
Iglesias MT, et al.
Journal of Food Engineering, 62(3), 215-223 (2004)
Peter M Kopittke et al.
Frontiers in plant science, 8, 1377-1377 (2017-08-22)
In wheat (Triticum aestivum), it is commonly assumed that Al is detoxified by the release of organic anions into the rhizosphere, but it is also possible that detoxification occurs within the apoplast and symplast of the root itself. Using Al-resistant
Debra Mohnen
Current opinion in plant biology, 11(3), 266-277 (2008-05-20)
Pectin is structurally and functionally the most complex polysaccharide in plant cell walls. Pectin has functions in plant growth, morphology, development, and plant defense and also serves as a gelling and stabilizing polymer in diverse food and specialty products and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service