P9121
Prolactin Receptor/Fc Chimera human
recombinant, expressed in mouse NSO cells, ≥90% (SDS-PAGE), lyophilized powder, suitable for cell culture
Synonym(s):
Lactotrophin, Luteotropic Hormone (LTH), Luteotropin, Mamotrophin, PRL
About This Item
biological source
human
recombinant
expressed in mouse NSO cells
Assay
≥90% (SDS-PAGE)
form
lyophilized powder
mol wt
~66 kDa by SDS-PAGE (reducing)
calculated mol wt ~51 kDa
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... PRLR(5618)
Physical form
Analysis Note
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service