Skip to Content
Merck
All Photos(1)

Key Documents

P8666

Sigma-Aldrich

Pyridoxine hydrochloride

suitable for plant cell culture

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058008
PubChem Substance ID:

form

powder

mol wt

Mw 205.64 g/mol

technique(s)

cell culture | plant: suitable

color

white to off-white

mp

214-215 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, colorless to faintly yellow

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fei Du et al.
STAR protocols, 2(1), 100301-100301 (2021-02-09)
Cortical microtubules (CMTs) play pivotal roles during plant cell growth and division. The organization of CMTs undergoes important changes during different cellular and developmental processes. Here, we describe two methods for the visualization of CMT organization in plant cells using
Ilan Matok et al.
Journal of clinical pharmacology, 53(3), 334-338 (2013-02-28)
Although Diclectin (doxylamine/pyridoxine delayed-released combination) is widely used in Canada, its pharmacokinetics (PK) during pregnancy has never been described. The objective of this study was to compare the PK of doxylamine/pyridoxine delayed-released combination in pregnant versus nonpregnant women. The apparent
Sonia Fargue et al.
Biochimica et biophysica acta, 1832(10), 1776-1783 (2013-04-20)
Primary hyperoxaluria type 1 (PH1) is a rare hereditary calcium oxalate kidney stone disease caused by a deficiency of the liver-specific pyridoxal-phosphate-dependent peroxisomal enzyme alanine:glyoxylate aminotransferase (AGT). About one third of patients are responsive to pharmacological doses of pyridoxine (vitamin
Eduard Alexander Struys et al.
Pediatrics, 130(6), e1716-e1719 (2012-11-14)
α-Amino adipic semialdehyde (α-AASA) accumulates in body fluids from patients with pyridoxine-dependent epilepsy because of mutations in antiquitin (ALDH7A1) and serves as the biomarker for this condition. We have recently found that the urinary excretion of α-AASA was also increased
Liat Ashkenazi-Hoffnung et al.
The Israel Medical Association journal : IMAJ, 15(1), 23-26 (2013-03-15)
Diclectin (pyridoxine 10 mg and doxylamine 10 mg) has traditionally been used to treat nausea and vomiting of pregnancy (NVP); however, this drug is unavailable in many countries. To evaluate the efficacy and safety of a simple bi-daily treatment regimen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service