Skip to Content
Merck
All Photos(1)

Key Documents

P8247

Sigma-Aldrich

Anti-Potassium Channel KIR3.3 (GIRK3) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
R$6,729.00

R$6,729.00


Check Cart for Availability


Select a Size

Change View
200 μL
R$6,729.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$6,729.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, mouse, human

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ9(3765)
mouse ... Kcnj9(16524)
rat ... Kcnj9(116560)

Immunogen

GST fusion protein with sequence corresponding to residues 344-361 of rat KIR3.3 (GIRK3). The sequence is identical in mouse and human.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Joo Yi et al.
Systems biology in reproductive medicine, 57(6), 296-308 (2011-11-08)
The GIRK2-containing inward-rectifying K(+) ion channels have been implicated in mammalian spermatogenesis. While the Girk2 null mice are fertile, the male weaver transgenic mice carrying a gain-of-function mutation in the Girk2 gene are infertile. To establish the exact period of
Carlo Corona et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(27), 6102-6113 (2018-06-08)
Activating Transcription Factor 4 (ATF4) has been postulated as a key regulator of learning and memory. We previously reported that specific hippocampal ATF4 downregulation causes deficits in synaptic plasticity and memory and reduction of glutamatergic functionality. Here we extend our

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service