Skip to Content
Merck
All Photos(2)

Key Documents

P7857

Sigma-Aldrich

Anti-P2X1 Purinergic Receptor antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RX1(5023)
mouse ... P2rx1(18436)
rat ... P2rx1(25505)

Related Categories

Immunogen

synthetic peptide corresponding to amino acids 382-399 of rat P2X1 (with additional N-terminal cysteine and serine). The epitope is highly conserved in mouse and human.

Physical form

Lyophilized at ~0.6 mg/ml from a solution of phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek M Kendig et al.
American journal of physiology. Renal physiology, 310(9), F909-F922 (2016-01-30)
Type II diabetes is the most prevalent form of diabetes. One of the primary complications of diabetes that significantly affects quality of life is bladder dysfunction. Many studies on diabetic bladder dysfunction have been performed in models of type I
Daniel Giglio et al.
The Journal of pharmacology and experimental therapeutics, 365(2), 327-335 (2018-03-14)
Currently, we have assessed the neuronal control of the urinary bladder in radiation cystitis and whether interstitial cells contribute to the condition. Fourteen days after bladder irradiation (20 Gy), rats were sedated and the urinary bladder was cut into two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service