Skip to Content
Merck
All Photos(1)

Key Documents

P7840

Sigma-Aldrich

L-α-Phosphatidylethanolamine, distearoyl methoxypolyethylene glycol conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
R$2,799.00

R$2,799.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
R$2,799.00

About This Item

MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

R$2,799.00


Check Cart for Availability

Request a Bulk Order

Assay

>98% (TLC)

Quality Level

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

Application

For the modification of liposome surfaces and for influencing the survival of liposomes in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service