Skip to Content
Merck
All Photos(1)

Key Documents

P7425

Sigma-Aldrich

Tetraethylene glycol monodecyl ether

~97% (GC)

Synonym(s):

C10E4, Decyl tetraethylene glycol ether, Decyltetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)4OH
CAS Number:
Molecular Weight:
334.49
Beilstein:
4247848
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

Assay

~97% (GC)

mol wt

334.49 g/mol

bp

288-290 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C18H38O5/c1-2-3-4-5-6-7-8-9-11-20-13-15-22-17-18-23-16-14-21-12-10-19/h19H,2-18H2,1H3

InChI key

ASMWIUUCZFNLHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylene glycol monodecyl ether is a surfactant.

Application

Tetraethylene glycol monodecyl ether has been used in a study to assess anomalous thickness variation of the foam films stabilized by weak non-ionic surfactants. It has also been used in a study to investigate removal of vaporous naphthalene using polyoxyethylenated nonionic surfactants.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cosima Stubenrauch et al.
Langmuir : the ACS journal of surfaces and colloids, 28(25), 9206-9210 (2012-06-08)
We showed in a previous study that a water-nonionic surfactant system, where the surfactant is a 9:1 mixture of tetraethylene glycol monodecyl ether (C(10)E(4)) and pentaethylene glycol monodecyl ether (C(10)E(5)), forms a disconnected lamellar (L(α)) phase. Thus, the isotropic phase
Christopher Hill et al.
Langmuir : the ACS journal of surfaces and colloids, 36(48), 14829-14840 (2020-11-25)
The interfacial properties and water-in-CO2 (W/CO2) microemulsion (μE) formation with double- and novel triple-tail surfactants bearing trimethylsilyl (TMS) groups in the tails are investigated. Comparisons of these properties are made with those for analogous hydrocarbon (HC) and fluorocarbon (FC) tail
Hsiao-Lin Huang et al.
Journal of the Air & Waste Management Association (1995), 53(8), 983-991 (2003-08-29)
Previous research has demonstrated that an anionic surfactant can increase the solubility of the vapor phases of both naphthalene and sulfur dioxide in water. This study examines the feasibility of removing polycyclic aromatic hydrocarbons (PAHs) during gas absorption by adding
Vishnu Sresht et al.
Langmuir : the ACS journal of surfaces and colloids, 33(33), 8319-8329 (2017-07-28)
A molecular modeling approach is presented with a focus on quantitative predictions of the surface tension of aqueous surfactant solutions. The approach combines classical Molecular Dynamics (MD) simulations with a molecular-thermodynamic theory (MTT) [ Y. J. Nikas, S. Puvvada, D.
Anne-Gaëlle Fournial et al.
Langmuir : the ACS journal of surfaces and colloids, 23(23), 11443-11450 (2007-10-13)
The binary phase diagram of tetraethylene glycol decanoyl ester (C9COE4) was investigated in the micellar region by PGSE-NMR (pulse field gradient spin echo nuclear magnetic resonance) and in the lamellar liquid crystalline state by 2H NMR. Its behavior was compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service