Skip to Content
Merck
All Photos(1)

Key Documents

P7296

Sigma-Aldrich

Peptone from animal tissue

BioReagent, Type I, suitable for plant cell culture, from meat

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
10171502
eCl@ss:
42040102

biological source

animal (meat)

type

Type I

product line

BioReagent

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Peptone P7296 is a meat enzyme hydrolysate composed of peptides and amino acids used as a supplement to support micropropagation and protocorm growth in media such as Murashige and Skoog, Knudson C (KnC) medium and VW medium.

Other Notes

Enzymatic hydrolysate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guy Parc et al.
Plant cell reports, 26(2), 145-152 (2006-09-15)
Growth and genetic stability of Nicotiana tabacum L. callus were strongly improved by replacing the inorganic nitrogen and phosphorus of the Murashige and Skoog's medium by a soybean peptone and phytate, respectively. Cell proliferation after subcultivation on the modified medium
B Kanjilal et al.
Indian journal of experimental biology, 38(11), 1164-1167 (2001-06-09)
Technique for rapid mass propagation of Geodorum densiflorum (Lam)Schltr. has been developed by using thin sections of stems of in vitro regenerated plantlets as explant source. Thin sections of stems (1.0-1.5 mm) when cultured in modified liquid and semisolid Knudson
M Firoz Alam et al.
Methods in molecular biology (Clifton, N.J.), 589, 21-28 (2010-01-26)
For high frequency clonal propagation of Vanda teres, nodal segments are cultured on VW medium supplemented with 2% sucrose, 2 mg/L Kinetin, 0.5 mg/L NAA, 2 g/L peptone, 1 g/L activated charcoal and 2.2 g/L gelrite. The cultures are incubated
Pinaki Sinha et al.
Methods in molecular biology (Clifton, N.J.), 589, 77-85 (2010-01-26)
For high frequency regeneration of Phalaenopsis, young leaf segments are cultured on gelrite-gelled (1/2) MS medium supplemented with 2% sucrose, 2.0 mg/L BA, 0.5 mg/L NAA, 10% coconut water (CW), 2 g/L peptone and 1 g/L activated charcoal. Cultures are
J Roy et al.
Indian journal of experimental biology, 39(10), 1041-1047 (2002-03-09)
Effects of different nutrient solutions, organic supplements and plant growth regulators on in vitro seed germination and protocorm development of Geodorum densiflorum (Lam.) Schltr. were studied. Seed germination was very high (up to 96%) in all the basal media, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service