Skip to Content
Merck
All Photos(1)

Key Documents

P6656

Sigma-Aldrich

Promazine hydrochloride

Synonym(s):

10-(3-Dimethylaminopropyl)phenothiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Beilstein:
3753230
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

solid

originator

Baxter

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2Sc3ccccc13

InChI

1S/C17H20N2S.ClH/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)20-17-11-6-4-9-15(17)19;/h3-6,8-11H,7,12-13H2,1-2H3;1H

InChI key

JIVSXRLRGOICGA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Promazine hydrochloride (PMZ) belongs to the phenothiazine drug family. [1] Promazine is a D2 dopamine receptor antagonist and has antipsychotic and anticholinergic functionality.[2] It is used for pain management and is also used for treating hypersensitivity reactions.[3]

Application

Promazine hydrochloride has been used in conductivity and surface tension measurements[1] and for testing inhibition of feline coronavirus (FCoV) in Felis catus whole fetus-4 (fcwf-4) cells.[4]

Biochem/physiol Actions

D2 dopamine receptor antagonist; phenothiazine antipsychotic.

Features and Benefits

This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Daniel et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 9(4), 337-344 (1999-07-28)
The aim of the present study was to investigate a possible impact of the three selective serotonin reuptake inhibitors (SSRIs) fluoxetine, fluvoxamine and sertraline on the pharmacokinetics of promazine in a steady state in rats. Promazine was administered twice a
Synergistic antiviral effect of Galanthus nivalis agglutinin and nelfinavir against feline coronavirus.
Hsieh LE, et al.
Antiviral Research, 88(1), 25-30 (2010)
Promazine in the treatment of delusional parasitosis.
Cubala W, et al.
Psychiatria Danubina, 23(2.), 198-199 (2011)
Study of the interaction between promazine hydrochloride and surfactant (conventional/gemini) mixtures at different temperatures.
Rub MA, et al.
Journal of Solution Chemistry, 43(5), 930-949 (2014)
Determination of thiazinamium, promazine and promethazine in pharmaceutical formulations using a CZE method.
Lara FJ, et al.
Analytica Chimica Acta, 535(1-2), 101-108 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service