Skip to Content
Merck
All Photos(1)

Key Documents

P5148

Sigma-Aldrich

pGlu-Asn-Gly

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N4O6
CAS Number:
Molecular Weight:
300.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

NC(=O)CC(NC(=O)C1CCC(=O)N1)C(=O)NCC(O)=O

InChI

1S/C11H16N4O6/c12-7(16)3-6(10(20)13-4-9(18)19)15-11(21)5-1-2-8(17)14-5/h5-6H,1-4H2,(H2,12,16)(H,13,20)(H,14,17)(H,15,21)(H,18,19)

InChI key

HYMUIMPABXIACR-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P M Cummins et al.
Biochimica et biophysica acta, 1429(1), 1-17 (1999-01-27)
Pyroglutamyl peptidase can be classified as an omega peptidase which hydrolytically removes the amino terminal pyroglutamate (pGlu) residue from specific pyroglutamyl substrates. To date, three distinct forms of this enzyme have been identified in mammalian tissues. Type I is typically
P M Cummins et al.
The international journal of biochemistry & cell biology, 28(8), 883-893 (1996-08-01)
Pyroglutamyl aminopeptidase type-1 (PAP-I) is reported to be a soluble, broad specificity aminopeptidase, capable of removing the pyroglutamic acid (pGlu) residue from the amino terminus of pGlu-peptides (e.g. TRH, LHRH, neurotensin and bombesin). The central aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service