Skip to Content
Merck
All Photos(1)

Key Documents

P5125

Sigma-Aldrich

DL-Penicillamine

Synonym(s):

β,β-Dimethyl-DL-cysteine, DL-2-Amino-3-mercapto-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(SH)CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Beilstein:
2039817
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

mp

204-208 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(S)C(N)C(O)=O

InChI

1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8)

InChI key

VVNCNSJFMMFHPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngmi Jo et al.
Journal of natural products, 69(10), 1399-1403 (2006-10-28)
Six new phenylethanoid glucosides, ternstrosides A-F (1-6), a new kaempferol derivative (7), and eight known compounds were isolated from the fresh leaves of Ternstroemia japonica. The structures were elucidated by 1D and 2D NMR spectroscopic analyses. Compounds 1-7 showed potent
Yanxia Xu et al.
Bioorganic & medicinal chemistry letters, 21(6), 1754-1757 (2011-02-15)
By linking the mercapto groups with isopropyl and introducing L-amino acid into the 5-carboxyl of DMSA a class of novel 5-(1-carbonyl-L-amino-acid)-2,2- dimethyl-[1,3]dithiolane-4-carboxylic acids were prepared. Their in vivo activities were evaluated on lead loaded mice at the dose of 0.4
J I Levin et al.
Bioorganic & medicinal chemistry letters, 15(19), 4345-4349 (2005-08-09)
The SAR of a series of potent sulfonamide hydroxamate TACE inhibitors bearing a butynyloxy P1' group was explored. In particular, compound 5k has excellent in vitro potency against TACE enzyme and in cells, and oral activity in an in vivo
N André Sasaki et al.
Bioorganic & medicinal chemistry, 17(6), 2310-2320 (2009-03-06)
2,3-Diaminopropionic acid (Dap) and N-terminal Dap peptides have been found to inhibit in vitro protein-modifications by methylglyoxal (MG), one of the highly reactive alpha-dicarbonyl compounds. MG scavenging potency of the newly synthesized N-terminal Dap peptides is demonstrated by RP-HPLC, SDS-PAGE
Clara Rosso-Fernández et al.
Trials, 16, 102-102 (2015-04-16)
Ventilator-associated pneumonia (VAP) is one of the most common and severe hospital-adquired infections, and multidrugresistant gram-negative bacilli (MDR-GNB) constitute the main etiology in many countries. Inappropriate empiric antimicrobial treatment is associated with increased mortality. In this context, the empirical treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service