Skip to Content
Merck
All Photos(1)

Key Documents

P4474

Sigma-Aldrich

Phosphate buffered saline

liquid, pH 7.4, sterile-filtered, suitable for cell culture

Synonym(s):

Phosphate buffered saline, PBS

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
R$154.00
1 L
R$249.00

R$154.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 ML
R$154.00
1 L
R$249.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.25

R$154.00


In StockDetails


Request a Bulk Order

Product Name

Phosphate buffered saline, pH 7.4, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

pH

7.4

Looking for similar products? Visit Product Comparison Guide

General description

Phosphate-buffered saline (PBS) is a buffer solution used in biological research. It is a water-based salt solution containing sodium phosphate, sodium chloride and, in some formulations, it contains potassium chloride and potassium phosphate. The osmolality and ion concentrations of the solutions match those of the human body (isotonic) and are non-toxic to most cells.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Questions

1–3 of 3 Questions  
  1. What is the conductivity of this solution?

    1 answer
    1. Unfortunately, the conductivity is not tested for this product as this is not a specification.

      Helpful?

  2. What is the concentration (in mM) of this solution?

    1 answer
    1. This product is a multicomponent item. The concentration and molarity of each component is as follows:
      Potassium phosphate monobasic - 0.144 g/L or 1.058mM
      Sodium chloride - 9.0 g/L or 154mM
      Sodium phosphate dibasic dihydrate - 0.528 g/L or 2.97mM

      Helpful?

  3. What is the composition of P4474 Phosphate buffered saline pH 7.4, liquid, sterile-filtered, suitable for cell culture ?

    1 answer
    1. Product P4474 contains the following:
      - 0.144 g/L Potassium phosphate monobasic
      - 9.0 g/L Sodium chloride
      - 0.528 g/L Sodium phosphate dibasic dihydrate

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service