Skip to Content
Merck
All Photos(1)

Key Documents

P3880

Sigma-Aldrich

Protamine chloride from salmon

Grade V, Histone, free(Millon test)

Synonym(s):

Protamine Chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$4,193.00

R$4,193.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
5 G
R$4,193.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

R$4,193.00


Estimated to ship onJune 02, 2025


biological source

fish (salmon)

type

Grade V

form

solid

impurities

Histone, free (Millon test)

solubility

H2O: soluble, clear

storage temp.

−20°C

Application

Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service