P3733
Hamilton® Softgrip® fixed-volume pipette
volume 10 μL
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
description
At 100% volume (Accuracy within ±0.80%, Precision within ±0.40%)
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Hamilton 55019-03
volume
10 μL
color
purple
General description
Same ergonomic features as the Hamilton Softgrip adjustable volume pipette.
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
- Less discomfort in the hand and wrist as compared to other pipettes during prolonged use
- Multi-point color coding for easy volume identification
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Softgrip is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)