Skip to Content
Merck
All Photos(1)

Documents

P3636

Sigma-Aldrich

Pentobarbital

Synonym(s):

5-Ethyl-5-(1-methylbutyl)-2,4,6-trioxohexahydropyrimidine, Nembutal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O3
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12352200

drug control

USDEA Schedule II; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

SMILES string

CCCC(C)C1(CC)C(=O)NC(=O)NC1=O

InChI

1S/C11H18N2O3/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16)

InChI key

WEXRUCMBJFQVBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoe Okamura et al.
Neuropharmacology, 58(1), 166-172 (2009-07-07)
Neuropeptide S (NPS) and its cognate receptor were reported to mediate anxiolytic-like and arousal effects. NPS receptors are predominantly expressed in the brain, especially in limbic structures, including amygdala, olfactory nucleus, subiculum and retrosplenial cortex. In contrast, the NPS precursor
M Katayama et al.
Laboratory animals, 47(2), 122-126 (2013-04-09)
Pharmacokinetics of drugs may differ between small and large mammals (including humans); therefore, drug testing in animal models must be carefully designed. Sprague-Dawley rats were used in cardiac experiments, during which the lopinavir concentration in serum had to match human
Nassir Mansour et al.
Journal of neurosurgery. Pediatrics, 12(1), 37-43 (2013-05-07)
Barbiturates are widely used in the management of high intracranial pressure (ICP) caused by diffuse brain swelling. The cardiovascular, renal, and immunological side effects of these drugs limit them to last-line therapy. There are few published data regarding the role
Brandon H Gilroyed et al.
Journal of veterinary science, 14(1), 103-106 (2013-02-08)
In the event of an infectious disease outbreak in cattle, carcasses must be disposed of in a rapid and contained manner. This brief communication details injection of a barbiturate to euthanize cattle inoculated with Escherichia coli O157:H7 followed by carcass
Maria Pedro Guarino et al.
In vivo (Athens, Greece), 27(1), 127-132 (2012-12-15)
We compared the effects of two different anaesthetics, sodium pentobarbital (65 mg/kg) and ketamine (30 mg/kg) plus xylazine (4 mg/kg) (KX) on insulin sensitivity, fasting glycaemia, insulinaemia and free fatty acids (FFA). Four groups of Wistar rats were used: KX

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service