Skip to Content
Merck
All Photos(1)

Documents

P2738

Sigma-Aldrich

PPNDS

solid

Synonym(s):

Pyridoxal-5′-phosphate-6-(2′-naphthylazo-6′-nitro-4′,8′-disulfonate) tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11N4O14PS2 · Na4
Molecular Weight:
694.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

dark red

solubility

H2O: 38 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[H]C(=O)c1c(O)c(C)nc(\N=N\c2cc(c3cc(cc(c3c2)S([O-])(=O)=O)[N+]([O-])=O)S([O-])(=O)=O)c1COP([O-])([O-])=O

Gene Information

human ... P2RX1(5023)

Biochem/physiol Actions

P2X1 Purinoreceptor antagonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lindström et al.
European journal of pharmacology, 387(1), 19-25 (2000-01-14)
ECL cells co-secrete histamine and pancreastatin, a chromogranin A-derived peptide, in response to gastrin. The aim of the study was to explore possible ways to deplete ECL cells of histamine without affecting pancreastatin and to examine how histamine depletion affects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service