Skip to Content
Merck
All Photos(2)

Key Documents

P233

Sigma-Aldrich

PD 168,077 maleate salt

powder

Synonym(s):

N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$3,239.00

R$3,239.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$3,239.00

About This Item

Empirical Formula (Hill Notation):
C20H22N4O · C4H4O4
CAS Number:
Molecular Weight:
450.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$3,239.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

form

powder

color

white

solubility

DMSO: >10 mg/mL

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.Cc1cccc(c1)C(=O)NCN2CCN(CC2)c3ccccc3C#N

InChI

1S/C20H22N4O.C4H4O4/c1-16-5-4-7-17(13-16)20(25)22-15-23-9-11-24(12-10-23)19-8-3-2-6-18(19)14-21;5-3(6)1-2-4(7)8/h2-8,13H,9-12,15H2,1H3,(H,22,25);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

NAEUGRPISCANHO-BTJKTKAUSA-N

Gene Information

human ... DRD4(1815)

Biochem/physiol Actions

Selective D4 dopamine receptor agonist.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Substituted [(4-phenylpiperazinyl)-methyl]benzamides: selective dopamine D4 agonists.
S A Glase et al.
Journal of medicinal chemistry, 40(12), 1771-1772 (1997-06-06)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service