Skip to Content
Merck
All Photos(1)

Key Documents

P2279

Sigma-Aldrich

Psilocin

Synonym(s):

4-Hydroxy-N,N-dimethyltryptamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule I; Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

storage temp.

−20°C

SMILES string

CN(C)CCc1c[nH]c2cccc(O)c12

InChI

1S/C12H16N2O/c1-14(2)7-6-9-8-13-10-4-3-5-11(15)12(9)10/h3-5,8,13,15H,6-7H2,1-2H3

InChI key

SPCIYGNTAMCTRO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hallucinogen originally isolated from the mushroom, Psilocybe mexicana, non-selective serotonin receptor agonist.
Hallucinogen; non-selective serotonin receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard Sard et al.
Bioorganic & medicinal chemistry letters, 15(20), 4555-4559 (2005-08-03)
An SAR study of psilocybin and psilocin derivatives reveals that 1-methylpsilocin is a selective agonist at the h5-HT(2C) receptor. The corresponding phosphate derivative, 1-methylpsilocybin, shows efficacy in an animal model for obsessive-compulsive disorder, as does 4-fluoro-N,N-dimethyltryptamine. These results suggest a
Kimberly G Nugent et al.
Forensic science international, 140(2-3), 147-157 (2004-03-24)
Hallucinogenic fungi synthesize two controlled substances, psilocin and psilocybin. Possession of the fungal species that contain these compounds is a criminal offence in North America. Some related species that are morphologically similar, do not contain the controlled substances. Therefore, unambiguous
Tooru Kamata et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 796(2), 421-427 (2003-10-29)
In order to develop a sensitive and reliable analytical method for psilocin (PC) in urine samples, the hydrolysis conditions including the acid, alkaline and enzymatic hydrolyses have been investigated by monitoring not only PC but also psilocin glucuronide (PCG) by
A Koçak et al.
Forensic science international, 195(1-3), 36-41 (2009-12-09)
This paper compares the use of ATR and transflection spectroscopic techniques for the qualitative analysis of psilocin extracted from hallucinogenic mushrooms and control spiked mushrooms. Both techniques gave comparable results and agreed with prior GC/MS analysis of the actual case
N Almaula et al.
Molecular pharmacology, 50(1), 34-42 (1996-07-01)
An important determinant of the neurobehavioral responses induced by a drug is its relative receptor selectivity. The molecular basis of ligand selectivity of hallucinogenic and nonhallucinogenic compounds of varying structural classes for the human 5-hydroxytryptamine (5-HT)2A and 5-HT2C receptors was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service