Skip to Content
Merck
All Photos(1)

Key Documents

P0435

Sigma-Aldrich

Pardaxin

≥97% (HPLC)

Synonym(s):

[Gly31]-Pardaxin P1, Gly-Phe-Phe-Ala-Leu-Ile-Pro-Lys-Ile-Ile-Ser-Ser-Pro-Leu-Phe-Lys-Thr-Leu-Leu-Ser-Ala-Val-Gly-Ser-Ala-Leu-Ser-Ser-Ser-Gly-Gly-Gln-Glu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C154H248N36O45
CAS Number:
Molecular Weight:
3323.83
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3ccccc3)NC(=O)CN)[C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)N4CCC[C@H]4C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)NCC(=O)NCC(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(O)=O

Biochem/physiol Actions

Induces neurotransmitter release

Other Notes

Originally isolated from Pardachirus marmoratus (Moses sole)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Shai et al.
FEBS letters, 242(1), 161-166 (1988-12-19)
Pardaxin, an amphipathic polypeptide secreted by the Red Sea flatfish Pardachirus marmoratus whose sequence is NH2-G-F-F-A-L-I-P-K-I-I-S-S-P-L-F-K-T-L-L-S-A-V-G-S-A-L-S-S-S-G-G-Q-E, was synthesized by the solid-phase method. The structure was verified by sequencing. The synthetic polypeptide changed the resistance of lipid bilayers by forming pores.
S A Thompson et al.
Science (New York, N.Y.), 233(4761), 341-343 (1986-07-18)
Three ichthyotoxic peptides, pardaxins P-I to P-3, have been isolated from the defense secretion of the sole Pardachirus pavoninus. Pavoninins, the steroid glycosides with shark-repelling ability, had previously been isolated therefrom. Each pardaxin consists of 33 amino acid residues having

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service