Skip to Content
Merck
All Photos(2)

Key Documents

P0117

Sigma-Aldrich

Monoclonal Anti-PIAS3 antibody produced in mouse

~2 mg/mL, clone PIA3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Protein Inhibitor of Activated STAT 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PIA3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~68 kDa (additional band at 44 kDa may be present)

species reactivity

chicken, mouse, canine, human, bovine

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 2-4 μg/mL using HepG2 total cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIAS3(10401)
mouse ... Pias3(229615)

General description

Protein inhibitor of activated STAT protein 3 (PIAS3) is an inhibitor of activated STAT3. The gene encodes for a 619-amino acid protein and has a molecular mass of 68 kDa. PIAS3 shares 55% identity with PIAS1 and PIASX proteins and 39% identity with PIASY. PIAS3 is widely expressed in various human tissues.

Immunogen

synthetic peptide corresponding to amino acids 591-604 of human PIAS3.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Immunohistochemistry (1 paper)
Monoclonal Anti-PIAS3 antibody produced in mouse has been used in:
  • enzyme linked immuno sorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

Protein inhibitor of activated STAT protein 3 (PIAS3) and other members of this family can act as small ubiquitin-like modifier (SUMO) E3 ligases and interfere with the transcription activity of many transcription factors. PIAS3 binds to microphthalmia-associated transcription factor (MITF), a key DNA-binding protein. It blocks MITF DNA-binding activity and co-expression of MITF. PIAS3 in NIH 3T3 cells inhibits MITF-driven transcription activity.

Target description

PIAS3 encodes a member of the PIAS [protein inhibitor of activated STAT (signal transducer and activator of transcription)] family of transcriptional modulators. The protein functions as a SUMO (small ubiquitin-like modifier)-E3 ligase which catalyzes the

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PIAS3 may represent a potential biomarker for diagnosis and therapeutic of human colorectal cancer
Li H, et al.
Medical Hypotheses, 81(6), 1151-1154 (2013)
A new role for the STAT3 inhibitor, PIAS3 a repressor of microphthalmia transcription factor
Levy C, et al.
The Journal of Biological Chemistry, 277(3), 1962-1966 (2002)
PIAS proteins and transcriptional regulation?more than just SUMO E3 ligases?
Sharrocks AD
Genes & Development, 20(7), 754-758 (2006)
Zohar Yagil et al.
Journal of immunology (Baltimore, Md. : 1950), 182(4), 2168-2175 (2009-02-10)
Protein inhibitor of activated STAT3 (PIAS3) functions in vivo as a key molecule in suppressing the transcriptional activity of both microphthalmia transcription factor (MITF) and STAT3, two transcription factors that play a major role in the development, phenotypic expression, and
Akishi Onishi et al.
Nature neuroscience, 13(9), 1059-1065 (2010-08-24)
Selective expression of retinal cone opsin genes is essential for color vision, but the mechanism mediating this process is poorly understood. Both vertebrate rod and medium wavelength-sensitive (M) cone photoreceptors differentiate by repression of a short wavelength-sensitive (S) cone differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service