Skip to Content
Merck
All Photos(1)

Key Documents

O5501

Sigma-Aldrich

Oxyphenonium bromide

Synonym(s):

2-([Cyclohexylhydroxyphenylacetyl]oxy)-N,N-diethyl-N-methylethanaminium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H34NO3Br
CAS Number:
Molecular Weight:
428.40
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

cationic

Assay

>98% (TLC)

mol wt

428.40 g/mol

SMILES string

Br.CC[N](C)(CC)CCOC(=O)C(O)(C1CCCCC1)c2ccccc2

InChI

1S/C21H34NO3.BrH/c1-4-22(3,5-2)16-17-25-20(23)21(24,18-12-8-6-9-13-18)19-14-10-7-11-15-19;/h6,8-9,12-13,19,24H,4-5,7,10-11,14-17H2,1-3H3;1H

InChI key

JKJFWDDPBNHJPV-UHFFFAOYSA-N

Application

Oxyphenonium bromide has been used in a study to assess masking mechanisms of bitter taste of drugs. It has also been used in a study to investigate stoichiometric and microenvironmental effects on hydrolysis of propantheline in cyclodextrin solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marian Turcani et al.
Scientific reports, 9(1), 2586-2586 (2019-02-24)
Successful adaptation to passive hyperthermia requires continual adjustment of circulation, which is mediated mainly by the autonomic nervous system. The goal of this study was to explore the alterations in spontaneous cardiovagal baroreflex sensitivity (BRS) during exposure to a hot
Alaa El-Gindy et al.
Journal of AOAC International, 90(5), 1250-1257 (2007-10-25)
A high-performance liquid chromatographic (HPLC) method was developed for determination of oxyphenonium bromide (OX) and its degradation product. The method was based on the HPLC separation of OX from its degradation product, using a cyanopropyl column at ambient temperature with
Noriaki Funasaki et al.
Chemical & pharmaceutical bulletin, 54(8), 1155-1161 (2006-08-02)
The masking mechanisms of the bitter taste of propantheline bromide (PB) and oxyphenonium (OB) bromide by native and modified cyclodextrins, saccharides, surfactants, organic acids, nonionic and anionic polymers, and other compounds were investigated with ion selective electrodes. The intensity of
Galina I Nezhinskaya et al.
Life sciences, 80(24-25), 2342-2346 (2007-05-15)
Cholinergic drugs can modulate anaphylactic shock and change lymphocyte functions. Plasma proteins modulate effects of muscarinic antagonists during anaphylactic shock. The present investigation was carried out to study the antianaphylactic activity of methacine (antagonist at muscarinic receptors) in combination with
[Use of endoscopic drug block of the vagus nerve and cryotherapy in the treatment of peptic ulcer].
V A Grushko et al.
Klinichna khirurhiia, (8)(8), 12-13 (1996-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service