Skip to Content
Merck
All Photos(1)

Key Documents

O3000

Sigma-Aldrich

Orotic acid sodium salt

Synonym(s):

6-Carboxy-2,4-dihydroxypyrimidine, Uracil-6-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
C5H3N2O4Na
CAS Number:
Molecular Weight:
178.08
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

solubility

1 M NaOH: 50 mg/mL, clear to very slightly hazy

SMILES string

[Na+].Oc1cc(nc(O)n1)C([O-])=O

InChI

1S/C5H4N2O4.Na/c8-3-1-2(4(9)10)6-5(11)7-3;/h1H,(H,9,10)(H2,6,7,8,11);/q;+1/p-1

InChI key

ZTHMWDSJKLNDTE-UHFFFAOYSA-M

comparable product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service