Skip to Content
Merck
All Photos(1)

Key Documents

N9505

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide–agarose

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

form

lyophilized powder

extent of labeling

1-5 μmol per mL

matrix activation

cyanogen bromide

matrix attachment

N-6

matrix spacer

11 atoms

storage temp.

−20°C

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service