Skip to Content
Merck
All Photos(1)

Key Documents

N9161

Sigma-Aldrich

Notch-2/Fc Chimera from rat

recombinant, expressed in NSO cells, ≥90% (SDS-PAGE), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

biological source

rat

Quality Level

recombinant

expressed in NSO cells

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

mol wt

105-115 kDa by SDS-PAGE (reducing)
calculated mol wt 76.5 kDa

impurities

endotoxin, tested

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

Gene Information

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to bind Jagged-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Weinmaster et al.
Development (Cambridge, England), 116(4), 931-941 (1992-12-01)
Notch is a cell surface receptor that mediates a wide variety of cellular interactions that specify cell fate during Drosophila development. Recently, homologs of Drosophila Notch have been isolated from Xenopus, human and rat, and the expression patterns of these
G Weinmaster
Current opinion in genetics & development, 10(4), 363-369 (2000-07-13)
The Notch signaling pathway functions in a wide variety of processes that regulate tissue patterning and morphogenesis in developing vertebrates and invertebrates. Research on the mechanism of ligand-induced Notch signal transduction has revealed a novel and essential element in the

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service