Skip to Content
Merck
All Photos(1)

Documents

N8143

Sigma-Aldrich

Neocarradodecaose 41,43,45,47,49,411-hexasulfate hexasodium salt

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C72H104O73S6Na6
CAS Number:
Molecular Weight:
2467.88
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥95%

form

solid

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].OC[C@H]1O[C@@H](O)[C@H](O)[C@@H](O[C@H]2O[C@@H]3CO[C@H]([C@H]2O)[C@H]3O[C@@H]4O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]5O[C@@H]6CO[C@H]([C@H]5O)[C@H]6O[C@@H]7O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]8O[C@@H]9CO[C@H]([C@H]8O)[C@H]9O[C@@H]%10O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]%11O[C@@H]%12CO[C@H]([C@H]%11O)[C@H]%12O[C@@H]%13O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]%14O[C@@H]%15CO[C@H]([C@H]%14O)[C@H]%15O[C@@H]%16O[C@H](CO)[C@H](OS([O-])(=O)=O)[C@H](O[C@H]%17O[C@@H]%18CO[C@@H]([C@H]%18O)[C@H]%17O)[C@H]%16O)[C@H]%13O)[C@H]%10O)[C@H]7O)[C@H]4O)[C@H]1OS([O-])(=O)=O

InChI

1S/C72H110O73S6.6Na/c73-1-13-43(140-146(93,94)95)55(26(80)61(92)117-13)134-63-28(82)50-38(20(124-63)8-112-50)129-69-34(88)57(45(15(3-75)119-69)142-148(99,100)101)136-65-30(84)52-40(22(126-65)10-114-52)131-71-36(90)59(47(17(5-77)121-71)144-150(105,106)107)138-67-32(86)54-42(24(128-67)12-116-54)133-72-37(91)60(48(18(6-78)122-72)145-151(108,109)110)139-66-31(85)53-41(23(127-66)11-115-53)132-70-35(89)58(46(16(4-76)120-70)143-149(102,103)104)137-64-29(83)51-39(21(125-64)9-113-51)130-68-33(87)56(44(14(2-74)118-68)141-147(96,97)98)135-62-27(81)49-25(79)19(123-62)7-111-49;;;;;;/h13-92H,1-12H2,(H,93,94,95)(H,96,97,98)(H,99,100,101)(H,102,103,104)(H,105,106,107)(H,108,109,110);;;;;;/q;6*+1/p-6/t13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25+,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38+,39+,40+,41+,42+,43+,44+,45+,46+,47+,48+,49+,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68+,69+,70+,71+,72+;;;;;;/m1....../s1

InChI key

YWSHUIVBCLNTEM-GGOYDXLKSA-H

Application

Neocarradodecaose has been used in a study to assess ionization and mass spectrometry of sulfated neocarrabiose oligosaccharides. It has also been used in a study to investigate ionic liquid matrixes optimized for MALDI-MS.

Other Notes

Mixed anomers

Preparation Note

Prepared enzymatically from κ-carrageenan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Fukuyama et al.
Analytical chemistry, 80(6), 2171-2179 (2008-02-16)
1,1,3,3-tetramethylguanidium (TMG) salt of alpha-cyano-4-hydroxycinnamic acid (CHCA) (G(2)CHCA) was reported by Tatiana et al. as a useful ionic liquid matrix (ILM) for sulfated oligosaccharides to suppress the loss of sulfate groups. However, the report mainly referred to positive ion spectra
Yuko Fukuyama et al.
Carbohydrate research, 337(17), 1553-1562 (2002-09-28)
Several commercial sulfated neocarrabiose oligosaccharides were analyzed by matrix-assisted ultraviolet laser-desorption ionization time-of-flight mass spectrometry (UV-MALDI-TOF-MS). UV-MALDI-TOF-MS was carried out in the linear and reflectron modes and, as routine, in both the positive- and negative-ion modes. 2,5-Dihydroxybenzoic acid and nor-harmane
M W McLean et al.
European journal of biochemistry, 93(3), 553-558 (1979-02-01)
A kappa-carrageenase was isolated from the cell-free medium of cultured Pseudomonas carrageenovora. From dodecylsulphate/polyacrylamide gel electrophoresis, a single protein (identified as the kappa-carrageenase) was detected in the medium. Activity against nominal carrageenan types and inspection of the products indicate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service