Skip to Content
Merck
All Photos(1)

Documents

N7779

Sigma-Aldrich

Nojirimycin bisulfite

solid, ≥98% (TLC)

Synonym(s):

5-Amino-5-deoxyglucopyranose bisulfite, Nojirimycin 1-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO7S
CAS Number:
Molecular Weight:
243.23
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Nojirimycin bisulfite microbial, solid

biological source

Streptomyces sp.

Assay

≥98% (TLC)

form

solid

storage temp.

−20°C

SMILES string

OC[C@H]1N[C@H]([C@H](O)[C@@H](O)[C@@H]1O)S(O)(=O)=O

InChI

1S/C6H13NO7S/c8-1-2-3(9)4(10)5(11)6(7-2)15(12,13)14/h2-11H,1H2,(H,12,13,14)/t2-,3-,4+,5-,6+/m1/s1

InChI key

PLICPKOWHZITQE-DVKNGEFBSA-N

Application

Inhibitor of β-glucosidases with about the same spectrum as parent nojirimycin, but 3- to 4-fold lower activity.
Inhibitor of β-glucosidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Donsimoni et al.
Enzyme, 39(2), 78-89 (1988-01-01)
This work describes the purification of a beta-glucosidase (beta-D-glucoside-glucohydrolase EC 3.2.1.21) from the digestive juice of Helix pomatia and the study of the enzyme's active site by using different reversible and irreversible inhibitors. The catalytic constants of arylglycosides and their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service