Skip to Content
Merck
All Photos(1)

Key Documents

N6521

Sigma-Aldrich

Neuropeptide Y Fragment 13-36 porcine

≥97% (HPLC)

Synonym(s):

NPY 13-36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C135H209N41O36
CAS Number:
Molecular Weight:
2982.36
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)[C@H](CO)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H]5CCCN5)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc6ccc(O)cc6)C(N)=O

InChI

1S/C135H209N41O36/c1-15-68(9)105(129(210)172-98(60-102(137)184)124(205)166-93(54-67(7)8)126(207)175-106(69(10)16-2)130(211)176-107(73(14)178)131(212)162-87(26-21-51-152-135(145)146)113(194)161-88(43-45-101(136)183)117(198)159-85(24-19-49-150-133(141)142)114(195)163-90(108(138)189)55-74-27-35-79(179)36-28-74)174-127(208)96(58-77-33-41-82(182)42-34-77)169-123(204)97(59-78-62-147-64-153-78)170-116(197)86(25-20-50-151-134(143)144)160-120(201)92(53-66(5)6)164-111(192)72(13)156-128(209)100(63-177)173-122(203)95(57-76-31-39-81(181)40-32-76)168-121(202)94(56-75-29-37-80(180)38-30-75)167-115(196)84(23-18-48-149-132(139)140)157-109(190)71(12)155-119(200)91(52-65(3)4)165-125(206)99(61-104(187)188)171-118(199)89(44-46-103(185)186)158-110(191)70(11)154-112(193)83-22-17-47-148-83/h27-42,62,64-73,83-100,105-107,148,177-182H,15-26,43-61,63H2,1-14H3,(H2,136,183)(H2,137,184)(H2,138,189)(H,147,153)(H,154,193)(H,155,200)(H,156,209)(H,157,190)(H,158,191)(H,159,198)(H,160,201)(H,161,194)(H,162,212)(H,163,195)(H,164,192)(H,165,206)(H,166,205)(H,167,196)(H,168,202)(H,169,204)(H,170,197)(H,171,199)(H,172,210)(H,173,203)(H,174,208)(H,175,207)(H,176,211)(H,185,186)(H,187,188)(H4,139,140,149)(H4,141,142,150)(H4,143,144,151)(H4,145,146,152)/t68-,69-,70-,71-,72-,73+,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,105-,106-,107-/m0/s1

InChI key

HEALDAASUSTTPJ-QGTLAHJGSA-N

Gene Information

pig ... NPY(397304)

Amino Acid Sequence

Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

Selective neuropeptide Y2 receptor agonist.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Nakajima et al.
Peptides, 19(2), 359-363 (1998-03-11)
In the present study, the effects of intracerebroventricular (ICV) NPY, [Leu31, Pro34]NPY and NPY13-36 have been evaluated with respect to anxiety in mice in the elevated plus maze. NPY had opposing effects on behavior, depending on the doses used. NPY
P J King et al.
Journal of neurochemistry, 73(2), 641-646 (1999-07-31)
Neuropeptide Y (NPY) is an important regulator of energy balance in mammals through its orexigenic, antithermogenic, and insulin secretagogue actions. We investigated the regulation of endogenous NPY release from rat hypothalamic slices by NPY receptor ligands and calcium channel antagonists.
B J McDermott et al.
European journal of pharmacology, 336(2-3), 257-265 (1998-01-24)
The aim of this study was to confirm the existence of and identify the receptor subtypes for neuropeptide Y that are present post-junctionally in myocardium. The effects of the selective agonists, [Leu31, Pro34] neuropeptide Y (neuropeptide Y Y1 receptors), neuropeptide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service