Skip to Content
Merck
All Photos(1)

Documents

N3536

Sigma-Aldrich

Neuraminidase 5X Reaction Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

liquid

Quality Level

storage temp.

2-8°C

Application

Neuraminidase is a glycoside hydrolase enzyme that is involved in the spread of the influenza virus. It is used to research potential drugs, such as neuraminidase inhibitors, to treat influenza. Neuraminidase mutations are studied for their implications on influenza drug resistance .

Biochem/physiol Actions

Neuraminidase cleaves the glycosidic linkages of neuraminic acids. Neuraminidase aids in the dispersment of the flu virus in mucosal secretions of the respiratory tract. It also prevents virions from self-aggregation .

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larisa V Gubareva
Virus research, 103(1-2), 199-203 (2004-05-28)
A wide use of inhibitors of influenza virus neuraminidase (NAIs) to control influenza in humans demands a better understanding of the mechanisms involved in the resistance emergence. In vitro studies demonstrate that both neuraminidase (NA) and hemagglutinin (HA) influence virus
In vitro passaging of a pandemic H1N1/09 virus selects for viruses with neuraminidase mutations conferring high-level resistance to oseltamivir and peramivir, but not to zanamivir
Jennifer L. McKimm-Breschkin, et al.
The Journal of Antimicrobial Chemotherapy, 10.1093, 150-150 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service