Skip to Content
Merck
All Photos(1)

Key Documents

MTCC2230

MTC Bio ScrewSeal Microcentrifuge Tubes

self-standing bottom, clear, capacity 2.0 mL, pack of 1000 ea (10 x bags 100 ea)

Synonym(s):

conical tube, plastic tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
Pricing and availability is not currently available.

material

blue high-density polyethylene cap
clear
polypropylene
screw top cap
self-standing bottom

sterility

sterile

feature

graduated

packaging

pack of 1000 ea (10 x bags 100 ea)

manufacturer/tradename

MTC Bio C2230

parameter

22,000 × g speed

capacity

2.0 mL

diam. × L

12.8 mm × 46 mm

Looking for similar products? Visit Product Comparison Guide

General description

Screw-cap microtubes, 2.0mL, with O-ring, sterile, molded graduations, caps attached, 100 per bag, 1000/case
ScrewSeal screw-cap microtubes are made from ultra-clear polypropylene, these screw cap microtubes include assembled caps with silicone O-rings that assure a leak-proof seal from -90°C to +121°C. Sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000×g.

Features and Benefits

  • Ultra-clear polypropylene
  • Molded or non-graduated options
  • Leak-proof O-ring seal
  • Light-blocking amber available
  • Sterile, RNase, DNase, Pyrogen free

Legal Information

MTC is a trademark of MTC Bio
ScrewSeal is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service