MBD0022
Inactivated Burkholderia pyrrocinia
Suitable for DNA extraction, PCR, sequencing and NGS, >10^8 bacteria/vial
Synonym(s):
PCR, Polymerase chain reaction
About This Item
Quality Level
form
powder
concentration
>10^8 bacteria/vial
technique(s)
DNA extraction: suitable
DNA sequencing: suitable
PCR: suitable
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Read here how to use our standards to ensure data integrity for your microbiome research.
Application
Physical form
Reconstitution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Turn to our Next-Gen Sequencing Oligos (NGSO) as a potential solution if you are experiencing high levels of adapter dimer formation with custom adapters sourced from other vendors.
DNA standards enhance metagenomics research integrity, offering precise species study and mixed community standards.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service