Skip to Content
Merck
All Photos(3)

Documents

MAK340

Sigma-Aldrich

Microsome Isolation Kit

sufficient for 50 isolation(s)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 50 isolation(s)

storage temp.

−20°C

General description

Microsomes are spherical vesicle-like structures formed from membrane fragments following homogenization and fractionation of eukaryotic cells. The microsomal subcellular fraction is prepared by differential centrifugation and consists primarily of membranes derived from the endoplasmic reticulum (ER) and Golgi apparatus. Microsomes isolated from liver tissue are used extensively in pharmaceutical development, toxicology and environmental science to study the metabolism of drugs, organic pollutants and other xenobiotic compounds by the cytochrome P450 monooxidase (CYP) enzyme superfamily. Microsomal preparations are an affordable and convenient in vitro system for assessing Phase I biotransformation reactions, as they contain all of the xenobiotic-metabolizing CYP isozymes and the membrane-bound flavoenzymes (such as NADPH P450-Reductase and cytochrome b5) required for function of the multicomponent P450 enzyme system. 

Suitability

Suitable for the isolation of Microsomes from mammalian glands and soft tissues such as liver, spleen, lungs etc.  or cultured eukaryotic cell lines such as HepG2 human hepatic carcinoma cells

Principle

The Microsome Isolation Kit enables preparation of active microsomes in about one hour, without the need for ultracentrifugation or sucrose gradient fractionation. The kit contains sufficient reagents for 50 isolation procedures, yielding microsomes from roughly 25 grams of tissue or cultured cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service