Skip to Content
Merck
All Photos(2)

Documents

MAK226

Sigma-Aldrich

Enrofloxacin Extraction Kit

greener alternative

Sufficient for 40 extractions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

cosmetics
food and beverages
sample preservation

greener alternative category

storage temp.

room temp

General description

Enrofloxacin is a fluoroquinolone antibiotic used for the treatment of pets and domestic animals and in fish farming. Minimum acceptable limits have been set for enrofloxacin residues in food samples. The enrofloxacin extraction kit enables the simultaneous separation of enrofloxacin from tissue, and the defatting of the sample during extraction in an accurate and quick manner.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product reduces solvent waste over traditional methods. Find details here.

Application

Enrofloxacin concentrations (ng/g) can be derived from biological specimens. Can be used for any biological food sample-shrimp, meat, liver, crab fish, etc. This kit enables the extraction of enrofloxacin for quantitation by ELISA.

Click on the following link for a video demonstration of how the kit works

Features and Benefits

The streamlined process bypasses the centrifugation, vortexing, pipetting and solvent addition steps associated with extracting and defatting the sample. The extracted enrofloxacin can then be detected and quantified by enzyme-linked immunosorbent assay (ELISA) followed by spectrophotometric analysis.

Suitability

This kit is suitable for the extraction of enroflaxacin from a variety of samples such as biological tissue and food samples.

Principle

The Enrofloxacin Extraction Kit enables the simultaneous separation of enrofloxacin from tissue, and the defatting of the sample during extraction in an accurate and quick manner. The streamlined process bypasses the centrifugation, vortexing, pipetting and solvent addition steps associated with extracting and defatting the sample. The extracted enrofloxacin can then be detected and quantified by enzyme-linked immunosorbent assay (ELISA) followed by spectrophotometric analysis.

Enrofloxacin is typically extracted using a single solvent system containing 80% methanol in buffer. In the standard procedure, methanol containing the enrofloxacin is separated from tissue by centrifugation for 10 minutes followed by manual pipetting. The extract is then dried and reconstituted into 1 mL of 8% methanol in sample dilution buffer. The extract is subsequently defatted by adding 1 mL hexane and centrifuging. A 50 μL aliquot is taken from the bottom layer containing the defatted enrofloxacin extract and used for analysis by ELISA and spectrophotometry.

The Enrofloxacin Extraction Kit shortcuts the extraction process by eliminating the need for centrifugation, manual pipetting, and post-extraction defatting with hexane or another lipophilic solvent. After adding 80% methanol to the sample, it can be mixed manually for 15 minutes or with sonication for 5 minutes and poured through the syringe containing a filter. The eluted solution containing the enrofloxacin extract is then reconstituted in 8% methanol and a 50 μL aliquot is used for analysis by ELISA and spectrophotometry.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service