Skip to Content
Merck
All Photos(3)

Key Documents

MAK091

Sigma-Aldrich

Hexokinase Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Hexokinase Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer; neurological disorders; hematological disorder

storage temp.

−20°C

General description

Hexokinases, which catalyze the ATP-dependent phosphorylation of aldo- and keto-hexoses to hexose-6-phosphate, catalyze the first step in a number of important biochemical pathways. In mammalian tissues, glucose is the predominant substrate for the hexokinases.[1] There are four isozymes of hexokinase in mammalian tissue, (HK-I, -II, -III, and -IV), which differ in their kinetic and regulatory properties as well as tissue distribution and cofactor use. The hexokinases catalyze the first step in most of the relevant glucose metabolism pathways. Alterations in hexokinase activity are associated with multiple disorders such as X-linked muscular dystrophy, hemolytic anermias, and cancer.[2]

Application

Hexokinase Colorimetric Assay Kit has been used to measure the activity of hexokinase.[3][4][5]

Suitability

Suitable for the measurement of Hexokinase activity, analysis of glucose metabolism and cell signaling, and screening anti-diabetic drugs in serum, animal tissues (liver, heart, kidney etc.), and cell culture (adherent or suspension cell).

Principle

The Hexokinase Colorimetric Assay kit provides a simple and direct procedure for measuring hexokinase activity in a variety of samples. Hexokinase activity is determined by a coupled enzyme assay, in which glucose is converted to glucose-6-phosphate by hexokinase, which is oxidized by glucose-6-phosphate dehydrogenase to form NADH. The resulting NADH reduces a colorless probe resulting in a colorimetric (450 nm) product proportional to the hexokinase activity present. One unit of HK is the amount of enzyme that will generate 1.0 μmole of NADH per minute at pH 8.0 at room temperature.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Endurance exercise causes adverse changes in some hematological and physio-biochemical indices in ponies under high altitude stress condition.
Vivek P, et al.
Indian J. Agri. Sci., 88(2), 222-228 (2018)
Md Wasim Khan et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(3), 678-687 (2018-12-14)
Hexokinase domain containing 1, a recently discovered putative fifth hexokinase, is hypothesized to play key roles in glucose metabolism. Specifically, during pregnancy in a recent genome wide association study (GWAS), a strong correlation between HKDC1 and 2-h plasma glucose in
Marybeth Anderson et al.
Oncotarget, 8(34), 56081-56094 (2016-06-01)
Pancreatic ductal adenocarcinoma (PDAC) is a KRAS-driven cancer with a high incidence of metastasis and an overall poor prognosis. Previous work in a genetically engineered mouse model of PDAC showed glucose metabolism to be important for maintaining tumor growth. Multiple
Crystal structures of an ATP-dependent hexokinase with broad substrate specificity from the hyperthermophilic archaeon Sulfolobus tokodaii.
Nishimasu H, et al.
The Journal of Biological Chemistry, 282(13), 9923-9931 (2007)
Endurance exercise causes adverse changes in some hematological and physio-biochemical indices in ponies under high altitude stress condition.
Vivek P, et al.
The Indian journal of animal sciences., 88(2), 222-228 (2018)

Protocols

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service