Skip to Content
Merck
All Photos(2)

Key Documents

MAK060

Sigma-Aldrich

Fumarate Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Fumarate Quantitative Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Fumarate is a tricarboxylic acid (TCA) cycle intermediate formed by the oxidation of succinate by succinate dehydrogenase. In the mammalian liver, Fumarate is also a product of the Urea cycle where its release in the cytosol leads to its conversion into malate and subsequently oxaloacetate while generating NADH in the cytosol. Deficiencies in fumarate hydratase are associated with increased cellular concentrations of fumarate and increased incidences of renal cell cancer. This is most likely due to fumarate-mediated upregulation of HIF1. The Fumarate Assay Kit provides a convenient tool for sensitive detection of the fumarate in a variety of samples.

Application

Fumarate Assay Kit has been used to determine the concentration of fumarate in samples.[1][2]

Suitability

Suitable for the detection of fumarate in a variety of samples including tissue, cells and serum

Principle

Fumarate is determined by an enzyme assay, which results in a colorimetric (450 nm) product, proportional to the fumarate present.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined effects of FH (E404D) and ACOX2 (R409H) cause metabolic defects in primary cardiac malignant tumor.
Zhou X, et al.
Cell Death & Disease, 5(1), 18-18 (2018)
Yijing Zhuang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 39(4), 1307-1316 (2016-09-09)
Motility is a feature of many pathogens that contributes to the migration and dispersion of the infectious agent. Whether gentamycin has a post-antibiotic effect (PAE) on the swarming and swimming motility of Escherichia coli (E. coli) remains unknown. In this
Melatonin alleviates adipose inflammation through elevating α-ketoglutarate and diverting adipose-derived exosomes to macrophages in mice.
Liu Z, et al.
Journal of Pineal Research, 64(1), e12455-e12455 (2018)
Xiangyu Zhou et al.
Cell death discovery, 4, 18-18 (2018-08-01)
Primary malignant cardiac tumors (PMCTs) are extremely rare. The apparent immunity of the heart to invasive cancer has attracted considerable interest given the continuously rising incidence of cancer in other organs. This study aims to determine the conditions that could
Ping Li et al.
International journal of molecular sciences, 23(3) (2022-02-16)
The FH gene encodes the fumarate hydratase of the Krebs cycle and functions as a homotetramer to catalyze the hydration of fumarate to malate. Mutations in FH result in uterine leiomyomas, a rare autosomal dominant inherited metabolic disease. However, how

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service