Skip to Content
Merck
All Photos(1)

Documents

M9630

Sigma-Aldrich

Met-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O3S
CAS Number:
Molecular Weight:
262.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CSCC[C@H](N)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C11H22N2O3S/c1-7(2)6-9(11(15)16)13-10(14)8(12)4-5-17-3/h7-9H,4-6,12H2,1-3H3,(H,13,14)(H,15,16)/t8-,9-/m0/s1

InChI key

PBOUVYGPDSARIS-IUCAKERBSA-N

Biochem/physiol Actions

Met-Leu is a dipeptide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Wei et al.
Analytical biochemistry, 250(1), 29-34 (1997-07-15)
A continuous assay for peptide deformylase has been developed using a formylated dipeptide, formyl-Met-Leu-p-nitroanilide, as substrate. Removal of the formyl group by a peptide deformylase renders the dipeptide product, which contains a free NH2 terminus, a substrate for an aminopeptidase
Jingjing Wang et al.
Atherosclerosis, 240(2), 335-344 (2015-04-16)
Mitochondrial dysfunction has been implicated in cardiovascular diseases. Elevation of serum retinol binding protein 4 (RBP4) in patients has been linked to cardiovascular disease. However, the role of RBP4 on mitochondrial oxidative stress and vascular oxidative damage is not well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service