Skip to Content
Merck
All Photos(1)

Documents

M8628

Sigma-Aldrich

α-Methyl-DL-histidine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3O2 · 2HCl
CAS Number:
Molecular Weight:
242.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98%

form

powder

color

white

application(s)

cell analysis

SMILES string

Cl.CC(N)(Cc1cnc[nH]1)C(O)=O

InChI

1S/C7H11N3O2.ClH/c1-7(8,6(11)12)2-5-3-9-4-10-5;/h3-4H,2,8H2,1H3,(H,9,10)(H,11,12);1H

InChI key

AABZGRFLFXCQEY-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather Francis et al.
Gut, 61(5), 753-764 (2011-08-30)
In several tumours the endogenous activity of histidine decarboxylase (HDC), the enzyme stimulating histamine synthesis, sustains the autocrine trophic effect of histamine on cancer progression. Cholangiocarcinoma is a biliary cancer with limited treatment options. Histamine interacts with four G-protein coupled
Olga Y Gasheva et al.
American journal of physiology. Gastrointestinal and liver physiology, 316(1), G217-G227 (2018-11-27)
This study aimed to establish mechanistic links between the prolonged intake of desloratadine, a common H1 receptor blocker (i.e., antihistamine), and development of obesity and metabolic syndrome. Male Sprague-Dawley rats were treated for 16 wk with desloratadine. We analyzed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service