Skip to Content
Merck
All Photos(1)

Key Documents

M8523

Sigma-Aldrich

Anti-β2-Microglobulin antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-B2MG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 1:5,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B2M(567)

General description

β-2-microglobulin is part of the human leukocyte antigen, where it represents the light chain. It is expressed on cell membranes and is made up of 99 amino acids. β-2-microglobulin is part of the immunoglobulin superfamily.
β2-microglobulin (B2M) is a non-glycosylated polypeptide. The protein is characterized with a single polypeptide chain, which is linked noncovalently to major histocompatibility complex (MHC) class I cell surface antigen. The gene coding for B2M is mapped to human chromosome 15q.

Immunogen

β2-microglobulin

Application

Anti-β2-Microglobulin antibody produced in rabbit has been used in immunoblottingand immunofluorescence.

Biochem/physiol Actions

β-2-microglobulin acts as a tumor marker and has been shown to be associated with dialysis-related amyloidosis.
β2-microglobulin (B2M) plays a vital role in cell survival, proliferation and metastasis in various types of cancer. Elevated concentration of β-2-microglobulin (B2M) can be considered as a prognostic marker for hemophagocytic lymphohistiocytosis (HLH). Overexpression of serum B2M is observed in hepatitis C virus (HCV) related chronic liver diseases, therefore it might act as a potential marker for the HCV disease progression towards cirrhosis and carcinoma. Missense mutation in the B2M gene results in familial hypercatabolic hypoproteinemia (FHH).

Quality

The antiserum has been treated to remove lipoprotens.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damien G Noone et al.
Kidney international, 90(1), 123-134 (2016-05-30)
Atypical hemolytic uremic syndrome and thrombotic thrombocytopenic purpura have traditionally been considered separate entities. Defects in the regulation of the complement alternative pathway occur in atypical hemolytic uremic syndrome, and defects in the cleavage of von Willebrand factor (VWF)-multimers arise
The prognostic significance of Beta2 microglobulin in patients with hemophagocytic lymphohistiocytosis
Jiang T, et al.
Disease Markers, 2016 (2016)
Expression of the neonatal Fc-receptor in placental-fetal endothelium and in cells of the placental immune system
Kiskova T, et al.
Placenta, 78, 36-43 (2019)
Annexin 2 regulates endothelial morphogenesis by controlling AKT activation and junctional integrity
Su SC, et al.
The Journal of Biological Chemistry, 285(52), 40624-40634 (2010)
Aruna P Ambagala et al.
Archives of virology, 158(5), 955-965 (2012-12-13)
Cynomolgus macaques are widely used as an animal model in biomedical research. We have established an immortalized cynomolgus macaque fibroblast cell line (MSF-T) by transducing primary dermal fibroblasts isolated from a 13-year-old male cynomolgus macaque with a retrovirus vector expressing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service