Skip to Content
Merck
All Photos(1)

Key Documents

M7011

Sigma-Aldrich

Mercury dibromofluorescein disodium salt

practical grade, crystalline

Synonym(s):

2′,7′-Dibromo-5′-(hydroxymercurio)fluorescein disodium salt, Merbromin, Mercurochrome

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br2HgNa2O6
CAS Number:
Molecular Weight:
750.65
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

grade

practical grade

form

crystalline

mp

≥300 °C

SMILES string

[Na+].[Na+].O[Hg]c1c([O-])c(Br)cc2c1OC3=CC(=O)C(Br)=CC3=C2c4ccccc4C([O-])=O

InChI

1S/C20H9Br2O5.Hg.2Na.H2O/c21-13-5-11-17(7-15(13)23)27-18-8-16(24)14(22)6-12(18)19(11)9-3-1-2-4-10(9)20(25)26;;;;/h1-7,24H,(H,25,26);;;;1H2/q;3*+1;/p-3

InChI key

SQFDQLBYJKFDDO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaux Mulatier et al.
Scientific reports, 9(1), 6664-6664 (2019-05-02)
Within mosquito vector populations, infectious mosquitoes are the ones completing the transmission of pathogens to susceptible hosts and they are, consequently, of great epidemiological interest. Mosquito infection by malaria parasites has been shown to affect several traits of mosquito physiology
J Contreras-Garduño et al.
Microbes and infection, 16(3), 261-267 (2013-12-03)
Immune priming is a new paradigm in innate immunity. However, most studies have focused on the benefits of priming (enhanced survival and parasite clearance after a second challenge), while little attention has been paid to the costs. In this study
G Nöhammer et al.
Histochemistry and cell biology, 107(5), 383-390 (1997-05-01)
Mercurochrom [2,7-dibromo-4-(hydroxymercuri)-fluorescein disodium salt] used for staining of protein thiols in addition binds to other groups of proteins. Experimental evidence is provided that mercurochrom bound to non-thiol groups forms a 1:1 adduct with protein (mixed) disulfides. The disulfide contents of
D O'Kane et al.
International journal of impotence research, 29(6), 225-228 (2017-07-28)
ED affects a significant proportion of males worldwide. With an ever-aging population the prevalence of ED is predicted to double in the next decade. Oral PDE-5 inhibitors are the first-line treatment for ED and have revolutionised its management. These agents
Local treatment of dry gangrene.
G Dreiplatz et al.
VASA. Zeitschrift fur Gefasskrankheiten, 31(4), 280-280 (2003-01-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service