Skip to Content
Merck
All Photos(1)

Documents

M6014

Sigma-Aldrich

Met-Leu-Phe acetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31N3O4S
CAS Number:
Molecular Weight:
409.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(N)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(O)=O

InChI

1S/C20H31N3O4S/c1-13(2)11-16(22-18(24)15(21)9-10-28-3)19(25)23-17(20(26)27)12-14-7-5-4-6-8-14/h4-8,13,15-17H,9-12,21H2,1-3H3,(H,22,24)(H,23,25)(H,26,27)

InChI key

CHDYFPCQVUOJEB-UHFFFAOYSA-N

Quality

Exhibits very weak chemotactic properties.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Banerjee et al.
Antimicrobial agents and chemotherapy, 42(2), 348-351 (1998-04-04)
The interaction of chemotactic peptide (e.g., fMet-Leu-Phe)-grafted liposomes with macrophages is noted to be rapid and specific. At a grafted peptide concentration of 100 nmol, internalization of the peptide-grafted liposomes by the macrophages is found to reach equilibrium in 30
Laurent Camous et al.
Blood, 117(4), 1340-1349 (2010-11-11)
Complement alternative pathway plays an important, but not clearly understood, role in neutrophil-mediated diseases. We here show that neutrophils themselves activate complement when stimulated by cytokines or coagulation-derived factors. In whole blood, tumor necrosis factor/formyl-methionyl-leucyl-phenylalanine or phorbol myristate acetate resulted
H L Nurcombe et al.
Annals of the rheumatic diseases, 48(1), 56-62 (1989-01-01)
Activated polymorphonuclear leucocytes (neutrophils) can generate both intracellular and extracellular luminol dependent chemiluminescence. As luminol dependent chemiluminescence largely measures the activity of the myeloperoxidase-H2O2 system, and as the extracellular activity of this enzyme may be responsible for the tissue damage
E S Buescher et al.
Cancer immunology, immunotherapy : CII, 37(1), 26-30 (1993-07-01)
During a phase I trial of interleukin-1 alpha (IL-1 alpha) in patients with ovarian carcinomas, the effects of this treatment on blood granulocyte respiratory burst and locomotive responses were examined. Differences in baseline granulocyte function in patients as well as
W Tao et al.
The Biochemical journal, 257(3), 633-637 (1989-02-01)
[3H]Arachidonic acid is released after stimulation of rabbit neutrophils with fMet-Leu-Phe or platelet-activating factor (PAF). The release is rapid and dose-dependent, and is inhibited in phorbol 12-myristate 13-acetate (PMA)-treated rabbit neutrophils. The protein kinase C (PKC) inhibitor 1-(5-isoquinoline-sulphonyl)-2-methylpiperazine (H-7) prevents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service