Skip to Content
Merck
All Photos(1)

Key Documents

M4537

Sigma-Aldrich

Mycolic acid from Mycobacterium tuberculosis (bovine strain)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

Mycobacterium tuberculosis (bovine strain)

description

mixture of homologous acids

form

powder

functional group

carboxylic acid

lipid type

unsaturated FAs

storage temp.

−20°C

Biochem/physiol Actions

Mycolic acids are long chain fatty acid containing molecules found in the cell walls of mycolata taxon bacteria. Mycolic acid from Mycobacterium tuberculosis (bovine strain) may be used to study mechanisms of resistance to antibiotic drug treatment and the toxicity of tuberculosis bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanghou Shui et al.
EMBO molecular medicine, 4(1), 27-37 (2011-12-08)
Mycolic acids are attractive diagnostic markers for tuberculosis (TB) infection because they are bacteria-derived, contain information about bacterial species, modulate host-pathogen interactions and are chemically inert. Here, we present a novel approach based on mass spectrometry. Quantification of specific precursor
Anna E Grzegorzewicz et al.
Nature chemical biology, 8(4), 334-341 (2012-02-22)
New chemotherapeutics active against multidrug-resistant Mycobacterium tuberculosis are urgently needed. We report on the identification of an adamantyl urea compound that shows potent bactericidal activity against M. tuberculosis and a unique mode of action, namely the abolition of the translocation
Damien J Montamat-Sicotte et al.
The Journal of clinical investigation, 121(6), 2493-2503 (2011-05-18)
Current tuberculosis (TB) vaccine strategies are largely aimed at activating conventional T cell responses to mycobacterial protein antigens. However, the lipid-rich cell wall of Mycobacterium tuberculosis (M. tuberculosis) is essential for pathogenicity and provides targets for unconventional T cell recognition.
Mervyn Beukes et al.
Chemistry and physics of lipids, 163(8), 800-808 (2010-09-30)
Cell wall mycolic acids (MA) from Mycobacterium tuberculosis (M.tb) are CD1b presented antigens that can be used to detect antibodies as surrogate markers of active TB, even in HIV coinfected patients. The use of the complex mixtures of natural MA
Asselineau, J.
The Bacterial Lipids,  -  (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service