Skip to Content
Merck
All Photos(2)

Key Documents

M3318

Sigma-Aldrich

Anti-MiRP1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Kcne2, Anti-MinK-related Peptide 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
rat
citations:
1
technique(s):
immunohistochemistry: suitable using rat ventricule
western blot: 1:200 using rat heart membranes and CHO transfected cells

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry: suitable using rat ventricule
western blot: 1:200 using rat heart membranes and CHO transfected cells

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KCNE2(9992)
mouse ... Kcne2(246133)
rat ... Kcne2(171138)

Immunogen

peptide corresponding to amino acid residues 88-107 of rat MiRP1. This epitope is highly homologous in mouse, human, and rabbit, respectively, 18/20, 16/20, and 15/20 residues identical.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn M Lamothe et al.
The Journal of biological chemistry, 291(39), 20387-20401 (2016-08-10)
The human ether-a-go-go-related gene (hERG) encodes the pore-forming subunit of the rapidly activating delayed rectifier potassium channel (IKr), which is important for cardiac repolarization. Dysfunction of hERG causes long QT syndrome and sudden death, which occur in patients with cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service