Skip to Content
Merck
All Photos(1)

Documents

M3275

Sigma-Aldrich

1-Methylxanthine

Synonym(s):

2,6-Dihydroxy-1-methylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
171507
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

≥300 °C

SMILES string

CN1C(=O)Nc2nc[nH]c2C1=O

InChI

1S/C6H6N4O2/c1-10-5(11)3-4(8-2-7-3)9-6(10)12/h2H,1H3,(H,7,8)(H,9,12)

InChI key

MVOYJPOZRLFTCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Feng Hsieh et al.
Bioorganic & medicinal chemistry, 15(10), 3450-3456 (2007-03-24)
Xanthine oxidase (XO) is a key enzyme which can catalyze xanthine to uric acid causing hyperuricemia in humans. By using the fractionation technique and inhibitory activity assay, an active compound that prevents XO from reacting with xanthine was isolated from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service